accessibility menu, dialog, popup

UserName

CAS RN 66323-49-7

NN(R)NHNO(S)(S)OH2NH2NHO

CAS RN 66323-49-7

IUPAC Name: 2-amino-9-[4-amino-5-(hydroxymethyl)oxolan-2-yl]-6,9-dihydro-3H-purin-6-one
Synonyms: 2-amino-9-[4-amino-5-(hydroxymethyl)oxolan-2-yl]-3H-purin-6-one
Molecular Weight (g/mol): 266.26
Molecular Formula: C10H14N6O3
InChi Key: HOQBPAWPBCAUGA-UHFFFAOYNA-N
SMILES: NC1CC(OC1CO)N1C=NC2=C1NC(N)=NC2=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results