accessibility menu, dialog, popup

UserName

CAS RN 6589-55-5

ClH3CNH2OH

CAS RN 6589-55-5

IUPAC Name: (2-hydroxy-2-phenylethyl)(methyl)azanium chloride
Synonyms: N-methylphenylethanolaminium chloride
Molecular Weight (g/mol): 187.67
Molecular Formula: C9H14ClNO
InChi Key: NPUGYWPZOLONFA-UHFFFAOYNA-N
SMILES: [Cl-].C[NH2+]CC(O)C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)