accessibility menu, dialog, popup

UserName

CAS RN 65414-74-6

HClNH2OHNH2O

CAS RN 65414-74-6

IUPAC Name: hydrogen 2-amino-3-hydroxypropanamide chloride
Synonyms: hydrogen 2-amino-3-hydroxypropanamide chloride
Molecular Weight (g/mol): 140.57
Molecular Formula: C3H9ClN2O2
InChi Key: VURWDDZIWBGXCK-UHFFFAOYNA-N
SMILES: [H+].[Cl-].NC(CO)C(N)=O
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)