accessibility menu, dialog, popup

UserName

CAS RN 654-11-5

H3COFOF

CAS RN 654-11-5

IUPAC Name: 3,5-difluoro-4-methoxybenzaldehyde
Synonyms: 3,5-difluoro-4-methoxybenzaldehyde
Molecular Weight (g/mol): 172.13
Molecular Formula: C8H6F2O2
InChi Key: KBSLGHDKSCGXKQ-UHFFFAOYSA-N
SMILES: COC1=C(F)C=C(C=O)C=C1F
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results