accessibility menu, dialog, popup

UserName

CAS RN 6515-58-8

OOBr

CAS RN 6515-58-8

IUPAC Name: 3-(bromomethyl)benzoate
Synonyms: 3-(bromomethyl)benzoate
Molecular Weight (g/mol): 214.04
Molecular Formula: C8H6BrO2
InChi Key: FTPAHNNMUYOHOB-UHFFFAOYSA-M
SMILES: [O-]C(=O)C1=CC=CC(CBr)=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results