CAS RN 639-14-5
CAS RN 639-14-5
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Gypsogenin, TRC
CAS: 639-14-5 Molecular Formula: C30H46O4 Molecular Weight (g/mol): 470.68 Synonym: Gypsophilasapogenin,Githagenin,Albsapogenin,Astrantiagenin D,(3β,4α)-3-Hydroxy-23-oxoolean-12-en-28-oic Acid; IUPAC Name: (4aS,6aR,6aS,6bR,8aR,9S,10S,12aR,14bS)-9-formyl-10-hydroxy-2,2,6a,6b,9,12a-hexamethyl-1,3,4,5,6,6a,7,8,8a,10,11,12,13,14b-tetradecahydropicene-4a-carboxylic acid SMILES: C[C@@]1(C2=O)[C@](CC2)([H])[C@@](CCC3=C4C=CC(O)=C3CC=C)([H])[C@]4([H])CC1