CAS RN 6290-03-5
IUPAC Name:
butane-1,3-diol
Synonyms:
1,3-butanediol butylene glycol 1,3 butylene glycol 1,3-butylene glycol 1,3-butylenglykol methyltrimethylene glycol β-butylene glycol caswell no. 128GG (R)-1,3-butylene glycol (r)-1,3-butylene glycol
Molecular Weight (g/mol):
90.12
Molecular Formula:
C4H10O2
InChi Key:
PUPZLCDOIYMWBV-UHFFFAOYNA-N
SMILES:
CC(O)CCO
1
–
4
of
4
results
1
(R)-(-)-1,3-Butanediol, 98%
CAS: 6290-03-5 MDL Number: MFCD00064277 ChEBI: CHEBI:52687
2
(R)-(-)-1,3-Butanediol, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
(R)-(-)-1,3-Butanediol, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
4
(R)-(-)-1,3-Butanediol, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.