CAS RN 624-51-1
CAS RN 624-51-1
IUPAC Name:
nonan-3-ol
Synonyms:
3-nonanol
Molecular Weight (g/mol):
144.26
Molecular Formula:
C9H20O
InChi Key:
GYSCXPVAKHVAAY-UHFFFAOYNA-N
SMILES:
CCCCCCC(O)CC
1
–
1
of
1
results
1
3-Nonanol, 98%
CAS: 624-51-1 Molecular Formula: C9H20O Molecular Weight (g/mol): 144.258 MDL Number: MFCD00014411 InChI Key: GYSCXPVAKHVAAY-UHFFFAOYSA-N Synonym: 3-nonanol,hexylethylcarbinol,ethylhexylcarbinol,3-nonyl alcohol,nonan-7-ol,ethyl hexyl carbinol,acmc-1b2h7 PubChem CID: 12216 IUPAC Name: nonan-3-ol SMILES: CCCCCCC(CC)O