accessibility menu, dialog, popup

UserName

CAS RN 623943-52-2

CH3SFFFFFNH2

CAS RN 623943-52-2

IUPAC Name: 2-methyl-5-(pentafluoro-λ⁶-sulfanyl)aniline
Synonyms: 2-methyl-5-(pentafluoro-λ⁶-sulfanyl)aniline
Molecular Weight (g/mol): 233.2
Molecular Formula: C7H8F5NS
InChi Key: IQTWYSPNHRBADI-UHFFFAOYSA-N
SMILES: CC1=C(N)C=C(C=C1)S(F)(F)(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results