accessibility menu, dialog, popup

UserName

CAS RN 623-50-7

H3COOOH

CAS RN 623-50-7

IUPAC Name: ethyl 2-hydroxyacetate
Synonyms: ethyl 2-hydroxyacetate
Molecular Weight (g/mol): 104.11
Molecular Formula: C4H8O3
InChi Key: ZANNOFHADGWOLI-UHFFFAOYSA-N
SMILES: CCOC(=O)CO
Molecular Weight (g/mol) 
  • (3)
  • (5)
Quantity 
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (2)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (3)
  • (5)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (3)
  • (5)