accessibility menu, dialog, popup

UserName

CAS RN 615-84-9

H3CNHOOHNCH3

CAS RN 615-84-9

IUPAC Name: N,N'-diethylethanediamide
Synonyms: N,N'-diethyl-oxalamide
Molecular Weight (g/mol): 144.17
Molecular Formula: C6H12N2O2
InChi Key: FFYAVOJIYAAUNX-UHFFFAOYSA-N
SMILES: CCNC(=O)C(=O)NCC

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results