accessibility menu, dialog, popup

UserName

CAS RN 6119-47-7

N(S)(R)(R)NH2CHOOH3CHHHClH2OH2O

CAS RN 6119-47-7

IUPAC Name: hydrogen (R)-[(2S,4R,5R)-5-ethenyl-1-azabicyclo[2.2.2]octan-2-yl](6-methoxyquinolin-4-yl)methanol dihydrate chloride
Synonyms: hydrogen QUIN dihydrate chloride
Molecular Weight (g/mol): 396.91
Molecular Formula: C20H29ClN2O4
InChi Key: MPQKYZPYCSTMEI-FXXIJDEQSA-N
SMILES: [H+].O.O.[Cl-].COC1=CC=C2N=CC=C([C@@H](O)[C@@H]3C[C@H]4CCN3C[C@@H]4C=C)C2=C1
Quantity 
  • (1)
  • (3)
  • (2)
  • (2)
  • (3)
  • (3)
Percent Purity 
  • (2)
  • (2)
  • (5)
Physical Form 
  • (3)
  • (3)
  • (2)
Color 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (4)
  • (2)
  • (1)
  • (3)
  • (2)
  • (3)
  • (2)
  • (1)
  • (3)
  • (2)
  • (2)
  • (3)
  • (3)
  • (2)
  • (2)
  • (5)
  • (3)
  • (3)
  • (2)
  • (2)
  • (2)
  • (2)