accessibility menu, dialog, popup

UserName

CAS RN 6105-74-4

H3CH3CNNH2

CAS RN 6105-74-4

IUPAC Name: 2-methyl-1-(piperidin-1-yl)propan-2-amine
Synonyms: 2-methyl-1-(piperidin-1-yl)propan-2-amine
Molecular Weight (g/mol): 156.27
Molecular Formula: C9H20N2
InChi Key: IMBNEKOMEDCTCL-UHFFFAOYSA-N
SMILES: CC(C)(N)CN1CCCCC1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results