accessibility menu, dialog, popup

UserName

CAS RN 59042-90-9

H3CNOH

CAS RN 59042-90-9

IUPAC Name: 1-(pyridin-2-yl)ethan-1-ol
Synonyms: 1-(pyridin-2-yl)ethanol
Molecular Weight (g/mol): 123.16
Molecular Formula: C7H9NO
InChi Key: PPHIIIRFJKDTLG-UHFFFAOYNA-N
SMILES: CC(O)C1=CC=CC=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results