CAS RN 58698-37-6
CAS RN 58698-37-6
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
(2S,3R)-2-Methyl-1,2,3,4-butanetetrol, TRC
CAS: 58698-37-6 Molecular Formula: C5 H12 O4 Molecular Weight (g/mol): 136.15 Synonym: [R-(R*,S*)]-2-Methyl-1,2,3,4-butanetetrol,2-C-Methyl-D-erythritol IUPAC Name: (2S,3R)-2-methylbutane-1,2,3,4-tetrol SMILES: C[C@](O)(CO)[C@H](O)CO