CAS RN 58652-20-3
CAS RN 58652-20-3
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Nomegestrol Acetate, TRC
CAS: 58652-20-3 Molecular Formula: C23 H30 O4 Molecular Weight (g/mol): 370.48 Synonym: Nomegestrol acetate,6-Methyl-3,20-dioxo-19-norpregna-4,6-dien-17-yl acetate IUPAC Name: [(8S,9S,10R,13S,14S,17R)-17-acetyl-6,13-dimethyl-3-oxo-1,2,8,9,10,11,12,14,15,16-decahydrocyclopenta[a]phenanthren-17-yl] acetate SMILES: CC(=O)O[C@@]1(CC[C@H]2[C@@H]3C=C(C)C4=CC(=O)CC[C@@H]4[C@H]3CC[C@]12C)C(=O)C