accessibility menu, dialog, popup

UserName

CAS RN 58543-16-1

H3CH2COOOHOHOOOHOHHOHOOOOHHOHOOHH3COOOOHOHOHOH

CAS RN 58543-16-1

IUPAC Name: 3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl 13-{[5-hydroxy-6-(hydroxymethyl)-3,4-bis({[3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy})oxan-2-yl]oxy}-5,9-dimethyl-14-methylidenetetracyclo[11.2.1.0¹,¹⁰.0⁴,⁹]hexadecane-5-carboxylate
Synonyms: rebaudioside A
Molecular Weight (g/mol): 967.02
Molecular Formula: C44H70O23
InChi Key: HELXLJCILKEWJH-UHFFFAOYNA-N
SMILES: CC12CCCC(C)(C1CCC13CC(=C)C(C1)(CCC23)OC1OC(CO)C(O)C(OC2OC(CO)C(O)C(O)C2O)C1OC1OC(CO)C(O)C(O)C1O)C(=O)OC1OC(CO)C(O)C(O)C1O
Grade 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results