accessibility menu, dialog, popup

UserName

CAS RN 58244-42-1

CH3ONOOBr

CAS RN 58244-42-1

IUPAC Name: 2-bromo-1-ethoxy-4-nitrobenzene
Synonyms: 2-bromo-1-ethoxy-4-nitrobenzene
Molecular Weight (g/mol): 246.06
Molecular Formula: C8H8BrNO3
InChi Key: DAJMJBJIPXARBJ-UHFFFAOYSA-N
SMILES: CCOC1=C(Br)C=C(C=C1)[N+]([O-])=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results