accessibility menu, dialog, popup

UserName

CAS RN 58123-77-6

OHOHOI

CAS RN 58123-77-6

IUPAC Name: 3-hydroxy-4-iodobenzoic acid
Synonyms: 3-hydroxy-4-iodobenzoic acid
Molecular Weight (g/mol): 264.02
Molecular Formula: C7H5IO3
InChi Key: UABBBWVTEWIIMN-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC(O)=C(I)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results