accessibility menu, dialog, popup

UserName

CAS RN 579476-63-4

CH3NBOHOH

CAS RN 579476-63-4

IUPAC Name: (2-methylpyridin-4-yl)boronic acid
Synonyms: 2-methylpyridin-4-ylboronic acid
Molecular Weight (g/mol): 136.95
Molecular Formula: C6H8BNO2
InChi Key: UFYBTLOLWSABAU-UHFFFAOYSA-N
SMILES: CC1=CC(=CC=N1)B(O)O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results