accessibility menu, dialog, popup

UserName

CAS RN 5767-36-2

ONNNHNH2N

CAS RN 5767-36-2

IUPAC Name: 4-(6-hydrazinylpyrimidin-4-yl)morpholine
Synonyms: 4-(6-hydrazinylpyrimidin-4-yl)morpholine
Molecular Weight (g/mol): 195.23
Molecular Formula: C8H13N5O
InChi Key: XWDNLTCJJYYYCI-UHFFFAOYSA-N
SMILES: NNC1=NC=NC(=C1)N1CCOCC1
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)