CAS RN 5743-04-4
CAS RN 5743-04-4
Cadmium acetate dihydrate, 98%
CAS: 5743-04-4 Molecular Formula: C4H10CdO6 Molecular Weight (g/mol): 266.53 MDL Number: MFCD00012449 InChI Key: AUIZLSZEDUYGDE-UHFFFAOYSA-L
Cadmium acetate dihydrate, 98%, pure
CAS: 5743-04-4 Molecular Formula: C4H6CdO4·2H2O Molecular Weight (g/mol): 266.52 InChI Key: AUIZLSZEDUYGDE-UHFFFAOYSA-L
Cadmium acetate dihydrate, 99.999% (metals basis)
CAS: 5743-04-4 Molecular Formula: C4H10CdO6 Molecular Weight (g/mol): 266.53 MDL Number: MFCD00012449 InChI Key: AUIZLSZEDUYGDE-UHFFFAOYSA-L
Cadmium acetate dihydrate, 98%, for analysis
CAS: 5743-04-4 Molecular Formula: C4H6CdO4·2H2O Molecular Weight (g/mol): 266.52 MDL Number: MFCD00150020 InChI Key: AUIZLSZEDUYGDE-UHFFFAOYSA-L
Cadmium Acetate Dihydrate, purum p.a., ≥98.0% (KT), Honeywell™
CAS: 5743-04-4 Molecular Formula: C4H10CdO6 Molecular Weight (g/mol): 266.53 MDL Number: MFCD00150020 InChI Key: AUIZLSZEDUYGDE-UHFFFAOYSA-L
Cadmium Acetate Dihydrate, 98%, Reagent grade, Honeywell™
CAS: 5743-04-4 Molecular Formula: C4H10CdO6 Molecular Weight (g/mol): 266.53 MDL Number: MFCD00150020 InChI Key: AUIZLSZEDUYGDE-UHFFFAOYSA-L