accessibility menu, dialog, popup

UserName

CAS RN 56-24-6

OHSnCH3CH3H3C

CAS RN 56-24-6

IUPAC Name: trimethylstannanol
Synonyms: trimethyltin hydroxide
Molecular Weight (g/mol): 182.84
Molecular Formula: C3H12OSn
InChi Key: RNRKJEKLVLXOLY-UHFFFAOYSA-N
SMILES: O.C[SnH](C)C
Quantity 
  • (1)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)