CAS RN 55774-32-8
CAS RN 55774-32-8
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
(7Z,9E)-Dodecadienyl Acetate, TRC
CAS: 55774-32-8 Molecular Formula: C14H24O2 Molecular Weight (g/mol): 224.34 Synonym: (E,Z)-7,9-Dodecadien-1-yl Acetate,(E,Z)-7,9-Dodecadienyl Acetate,(Z,E)-7,9-Dodecadienyl Acetate; SMILES: CC\C=C\C=C/CCCCCCOC(=O)C