accessibility menu, dialog, popup

UserName

CAS RN 5557-83-5

HClH3CNH2OO

CAS RN 5557-83-5

IUPAC Name: hydrogen benzyl 2-aminopropanoate chloride
Synonyms: hydrogen benzyl 2-aminopropanoate chloride
Molecular Weight (g/mol): 215.68
Molecular Formula: C10H14ClNO2
InChi Key: RLMHWGDKMJIEHH-UHFFFAOYNA-N
SMILES: [H+].[Cl-].CC(N)C(=O)OCC1=CC=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (1)