accessibility menu, dialog, popup

UserName

CAS RN 5486-84-0

CH3ONNOCH3HNO

CAS RN 5486-84-0

IUPAC Name: 4-benzamido-2,5-diethoxybenzene-1-diazonium
Synonyms: 4-benzamido-2,5-diethoxybenzenediazonium
Molecular Weight (g/mol): 312.35
Molecular Formula: C17H18N3O3
InChi Key: DKPBKNZQVUOHRQ-UHFFFAOYSA-O
SMILES: CCOC1=CC([N+]#N)=C(OCC)C=C1NC(=O)C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (1)
  • (3)
Quantity 
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (1)
  • (3)
  • (1)
  • (1)
  • (2)