accessibility menu, dialog, popup

UserName

CAS RN 547-66-0

MgOOOO

CAS RN 547-66-0

IUPAC Name: magnesium(2+) oxalate
Synonyms: magnesium oxalate
Molecular Weight (g/mol): 112.32
Molecular Formula: C2MgO4
InChi Key: UHNWOJJPXCYKCG-UHFFFAOYSA-L
SMILES: [Mg++].[O-]C(=O)C([O-])=O
Percent Purity 
  • (3)
Physical Form 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results