CAS RN 546141-08-6
CAS RN 546141-08-6
IUPAC Name:
3'-carbamoyl-[1,1'-biphenyl]-3-yl N-cyclohexylcarbamate
Synonyms:
3'-carbamoyl-[1,1'-biphenyl]-3-yl N-cyclohexylcarbamate
Molecular Weight (g/mol):
338.41
Molecular Formula:
C20H22N2O3
InChi Key:
ROFVXGGUISEHAM-UHFFFAOYSA-N
SMILES:
NC(=O)C1=CC=CC(=C1)C1=CC=CC(OC(=O)NC2CCCCC2)=C1
1
–
1
of
1
results
1
URB-597, MedChemExpress
MedChemExpress URB-597 (KDS-4103) is an orally bioavailable and selective FAAH inhibitor. URB-597 inhibits FAAH activity with an IC50s of approximately 5 nM in rat brain membranes, 0.5 nM in intact rat neurons, 3 nM in human liver microsomes. Antidepressant-like effects. Analgesic activity.