accessibility menu, dialog, popup

UserName

CAS RN 537-03-1

LaLaOOOOOOOOOOOO

CAS RN 537-03-1

IUPAC Name: dilanthanum(3+) trioxalate
Synonyms: dilanthanum(3+) trioxalate
Molecular Weight (g/mol): 541.87
Molecular Formula: C6La2O12
InChi Key: OXHNIMPTBAKYRS-UHFFFAOYSA-H
SMILES: [La+3].[La+3].[O-]C(=O)C([O-])=O.[O-]C(=O)C([O-])=O.[O-]C(=O)C([O-])=O
Grade 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results