accessibility menu, dialog, popup

UserName

CAS RN 5369-19-7

CH3H3CCH3H2N

CAS RN 5369-19-7

IUPAC Name: 3-tert-butylaniline
Synonyms: 3-tert-butylaniline
Molecular Weight (g/mol): 149.24
Molecular Formula: C10H15N
InChi Key: DPKTVUKEPNBABS-UHFFFAOYSA-N
SMILES: CC(C)(C)C1=CC=CC(N)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results