CAS RN 53527-42-7
CAS RN 53527-42-7
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Luteolin-3-O-beta-D-glucuronide, MedChemExpress
MedChemExpress Luteolin-3-O-beta-D-glucuronide is a luteolin glucosiduronic acid consisting of luteolin having a beta-D-glucosiduronic acid residue attached at the 3'-position.