accessibility menu, dialog, popup

UserName

CAS RN 53515-19-8

CH3H2NS

CAS RN 53515-19-8

IUPAC Name: 2-methylbenzene-1-carbothioamide
Synonyms: 2-methylbenzenecarbothioamide
Molecular Weight (g/mol): 151.23
Molecular Formula: C8H9NS
InChi Key: DEFUMLHLEJNWIO-UHFFFAOYSA-N
SMILES: CC1=CC=CC=C1C(N)=S

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results