CAS RN 52788-53-1
CAS RN 52788-53-1
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
Gadolinium(III) nitrate pentahydrate, 99.9%
CAS: 52788-53-1 Molecular Formula: GdN3O9·5H2O Molecular Weight (g/mol): 433.33 MDL Number: MFCD00149729