accessibility menu, dialog, popup

UserName

CAS RN 52182-15-7

H3COOHO

CAS RN 52182-15-7

IUPAC Name: ethyl 2-hydroxy-2,2-diphenylacetate
Synonyms: ethyl 2-hydroxy-2,2-diphenylacetate
Molecular Weight (g/mol): 256.3
Molecular Formula: C16H16O3
InChi Key: AIPVNQQMYPWQSX-UHFFFAOYSA-N
SMILES: CCOC(=O)C(O)(C1=CC=CC=C1)C1=CC=CC=C1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results