accessibility menu, dialog, popup

UserName

CAS RN 514-65-8

OHN

CAS RN 514-65-8

IUPAC Name: 1-{bicyclo[2.2.1]hept-5-en-2-yl}-1-phenyl-3-(piperidin-1-yl)propan-1-ol
Synonyms: biperiden akineton biperidene biperideno biperidenum akinophyl biperiden (product) biperiden (substance) biperiden preparation
Molecular Weight (g/mol): 311.47
Molecular Formula: C21H29NO
InChi Key: YSXKPIUOCJLQIE-UHFFFAOYNA-N
SMILES: OC(CCN1CCCCC1)(C1CC2CC1C=C2)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results