accessibility menu, dialog, popup

UserName

CAS RN 513-36-0

CH3H3CCl

CAS RN 513-36-0

IUPAC Name: 1-chloro-2-methylpropane
Synonyms: isobutyl chloride
Molecular Weight (g/mol): 92.57
Molecular Formula: C4H9Cl
InChi Key: QTBFPMKWQKYFLR-UHFFFAOYSA-N
SMILES: CC(C)CCl
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)