accessibility menu, dialog, popup

UserName

CAS RN 512-04-9

H3C(R)(R)OO(S)(S)(S)(S)OH(R)H3C(S)(S)H3C(R)(S)H3C

CAS RN 512-04-9

IUPAC Name: (1'S,2R,2'S,4'S,5R,7'S,8'R,9'S,12'S,13'R,16'S)-5,7',9',13'-tetramethyl-5'-oxaspiro[oxane-2,6'-pentacyclo[10.8.0.0²,⁹.0⁴,⁸.0¹³,¹⁸]icosan]-18'-en-16'-ol
Synonyms: diosgenin diosgenin (substance) 22α-spirost-5-en-3β-ol (25R)-spirost-5-en-3-β-ol nitogenin
Molecular Weight (g/mol): 414.63
Molecular Formula: C27H42O3
InChi Key: WQLVFSAGQJTQCK-VKROHFNGSA-N
SMILES: C[C@H]1[C@H]2[C@H](C[C@H]3[C@@H]4CC=C5C[C@@H](O)CC[C@]5(C)[C@H]4CC[C@]23C)O[C@]11CC[C@@H](C)CO1
Molecular Weight (g/mol) 
  • (3)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (1)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (3)