accessibility menu, dialog, popup

UserName

CAS RN 50907-31-8

ClNNHNNCl

CAS RN 50907-31-8

IUPAC Name: 5-(2,6-dichlorophenyl)-2H-1,2,3,4-tetrazole
Synonyms: 5-(2,6-dichlorophenyl)-2H-1,2,3,4-tetrazole
Molecular Weight (g/mol): 215.04
Molecular Formula: C7H4Cl2N4
InChi Key: RLXQNTWJPTUTCL-UHFFFAOYSA-N
SMILES: ClC1=CC=CC(Cl)=C1C1=NNN=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results