CAS RN 5053-43-0
CAS RN 5053-43-0
IUPAC Name:
2-methylpyrimidine
Synonyms:
pyrimidine, 2-methyl-
Molecular Weight (g/mol):
94.12
Molecular Formula:
C5H6N2
InChi Key:
LNJMHEJAYSYZKK-UHFFFAOYSA-N
SMILES:
CC1=NC=CC=N1
1
–
1
of
1
results
1
2-Methylpyrimidine, 98%
CAS: 5053-43-0 Molecular Formula: C5H6N2 Molecular Weight (g/mol): 94.11 InChI Key: LNJMHEJAYSYZKK-UHFFFAOYSA-N Synonym: pyrimidine, 2-methyl,2-methyl pyrimidine,pubchem21974,2-methyl-1,3-diazine,2-methylpyrimidine,pyrimidine, 2-methyl-6ci,7ci,8ci,9ci PubChem CID: 78748 IUPAC Name: 2-methylpyrimidine SMILES: CC1=NC=CC=N1