CAS RN 502-52-3
CAS RN 502-52-3
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Glycerol 1,3-Dipalmitate, TRC
CAS: 502-52-3 Molecular Formula: C35H68O5 Molecular Weight (g/mol): 568.91 Synonym: 1,3-Dipalmitin,1,3-Dipalmitoylglycerin,1,3-Dipalmitoylglycerol,Glycerol 1,3-Dihexadecanoate,Hexadecanoic Acid, 2-hydroxy-1,3-propanediyl Ester IUPAC Name: (3-hexadecanoyloxy-2-hydroxypropyl) hexadecanoate SMILES: CCCCCCCCCCCCCCCC(=O)OCC(O)COC(=O)CCCCCCCCCCCCCCC