accessibility menu, dialog, popup

UserName

CAS RN 499785-47-6

CH3NNHO

CAS RN 499785-47-6

IUPAC Name: (1-methyl-5-phenyl-1H-pyrazol-4-yl)methanol
Synonyms: (1-methyl-5-phenylpyrazol-4-yl)methanol
Molecular Weight (g/mol): 188.23
Molecular Formula: C11H12N2O
InChi Key: ZVGZXURABHDVPS-UHFFFAOYSA-N
SMILES: CN1N=CC(CO)=C1C1=CC=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results