accessibility menu, dialog, popup

UserName

CAS RN 49783-84-8

CH3NNH3COCl

CAS RN 49783-84-8

IUPAC Name: 1,5-dimethyl-1H-pyrazole-3-carbonyl chloride
Synonyms: 1,5-dimethylpyrazole-3-carbonyl chloride
Molecular Weight (g/mol): 158.59
Molecular Formula: C6H7ClN2O
InChi Key: BADBQYAILRGCBB-UHFFFAOYSA-N
SMILES: CN1N=C(C=C1C)C(Cl)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results