accessibility menu, dialog, popup

UserName

CAS RN 4894-75-1

O

CAS RN 4894-75-1

IUPAC Name: 4-phenylcyclohexan-1-one
Synonyms: cyclohexanone, 4-phenyl-
Molecular Weight (g/mol): 174.24
Molecular Formula: C12H14O
InChi Key: YKAYMASDSHFOGI-UHFFFAOYSA-N
SMILES: O=C1CCC(CC1)C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)