CAS RN 487020-03-1
CAS RN 487020-03-1
IUPAC Name:
N-{12-cyanoindolizino[2,3-b]quinoxalin-2-yl}thiophene-2-carboxamide
Synonyms:
N-{12-cyanoindolizino[2,3-b]quinoxalin-2-yl}thiophene-2-carboxamide
Molecular Weight (g/mol):
369.4
Molecular Formula:
C20H11N5OS
InChi Key:
BCSBXWKRZUPFHW-UHFFFAOYSA-N
SMILES:
O=C(NC1=CC2=C(C#N)C3=NC4=CC=CC=C4N=C3N2C=C1)C1=CC=CS1
1
–
1
of
1
results
1