accessibility menu, dialog, popup

UserName

CAS RN 4819-69-6

ClNOO

CAS RN 4819-69-6

IUPAC Name: 2-(4-chlorobut-2-yn-1-yl)-2,3-dihydro-1H-isoindole-1,3-dione
Synonyms: 2-(4-chlorobut-2-yn-1-yl)isoindole-1,3-dione
Molecular Weight (g/mol): 233.65
Molecular Formula: C12H8ClNO2
InChi Key: TXNDRPKNOXQAAO-UHFFFAOYSA-N
SMILES: ClCC#CCN1C(=O)C2=CC=CC=C2C1=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results