accessibility menu, dialog, popup

UserName

CAS RN 475-20-7

CH3H3CH3CCH2

CAS RN 475-20-7

IUPAC Name: 3,3,7-trimethyl-8-methylidenetricyclo[5.4.0.0²,⁹]undecane
Synonyms: longifolene
Molecular Weight (g/mol): 204.36
Molecular Formula: C15H24
InChi Key: PDSNLYSELAIEBU-UHFFFAOYNA-N
SMILES: CC12CCCC(C)(C)C3C(CCC13)C2=C
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results