accessibility menu, dialog, popup

UserName

CAS RN 4746-32-1

H3CHN

CAS RN 4746-32-1

IUPAC Name: N-hexylaniline
Synonyms: N-hexylaniline
Molecular Weight (g/mol): 177.29
Molecular Formula: C12H19N
InChi Key: OXHJCNSXYDSOFN-UHFFFAOYSA-N
SMILES: CCCCCCNC1=CC=CC=C1
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)