CAS RN 474-63-5
CAS RN 474-63-5
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Ostreasterol, TRC
CAS: 474-63-5 Molecular Formula: C28 H46 O Molecular Weight (g/mol): 398.66 Synonym: 24-Methylenecholesterol,NSC 232664,24-Methylene-cholesterol,24-Methylenecholest-5-en-3β-ol,(3β)-ergosta-5,24(28)-dien-3-ol,Ostreasterol IUPAC Name: (3S,8S,9S,10R,13R,14S,17R)-10,13-dimethyl-17-[(2R)-6-methyl-5-methylideneheptan-2-yl]-2,3,4,7,8,9,11,12,14,15,16,17-dodecahydro-1H-cyclopenta[a]phenanthren-3-ol SMILES: CC(C)C(=C)CC[C@@H](C)[C@H]1CC[C@H]2[C@@H]3CC=C4C[C@@H](O)CC[C@]4(C)[C@H]3CC[C@]12C