CAS RN 4670-05-7
CAS RN 4670-05-7
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Theaflavin, MedChemExpress
MedChemExpress Theaflavin is a suitable natural inhibitor against influenza A (H1N1) neuraminidase.