CAS RN 464-43-7
CAS RN 464-43-7
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
(+)-Borneol, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
(+)-Borneol, TRC
CAS: 464-43-7 Molecular Formula: C10 H18 O Molecular Weight (g/mol): 154.25 Synonym: Bicyclo[2.2.1]heptan-2-ol, 1,7,7-trimethyl-, (1R,2S,4R)-,Bicyclo[2.2.1]heptan-2-ol, 1,7,7-trimethyl-, (1R-endo)-,Borneol, (1R,2S,4R)-(+)- (8CI),(1R,2S,4R)-1,7,7-Trimethylbicyclo[2.2.1]heptan-2-ol,(+)-(2S)-Borneol,(+)-Borneol,(+)-endo-Borneol,(1R)-(+)-Borneol,(1R)-endo-Borneol,Borneol, (+)-,d-(+)-Borneol,d-Borneol IUPAC Name: (1R,3S,4R)-4,7,7-trimethylbicyclo[2.2.1]heptan-3-ol SMILES: CC1(C)[C@@H]2CC[C@@]1(C)[C@@H](O)C2